Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAPSA Recombinant Rabbit Monoclonal Antibody (PD01-30)
GREENER_CHOICE

Catalog No. FEMA550741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550741 Supplier Invitrogen™ Supplier No. MA550741
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Human lung carcinoma tissue, human kidney tissue. Predicted band size: 45 kDa Subcellular Location: Secreted.

Napsin 1 is an aspartic protease that is generally expressed in the lung, but also present in kidney and in metastatic carcinomas throughout the body. Napsin 1 processes surfactant protein B in healthy pulmonary and renal tissues, but is abnormally regulated when carcinogenesis occurs. Thus, many researchers have found napsin evaluation to be useful in identifying several cancers, and differentiating primary tumors from metastases. When combined with thyroid transcription factor analysis, napsin can be used as a specific biomarker for differentiation of carcinoma types. Napsin has also been associated with pulmonary alveolar proteinosis as well as in protein catabolism in renal tubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAPSA
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone PD01-30
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NAPSA
Gene Accession No. O96009
Gene Alias asp 4; ASP4; Aspartyl protease 4; KAP; Kdap; KDAP-1; kidney-derived aspartic protease-like protein; Nap; NAP1; NAPA; Napsa; napsin A aspartic peptidase; napsin-1; Napsin-A; pronapsin; pronapsin A; SNAPA; TA01/TA02
Gene Symbols NAPSA
Host Species Rabbit
Immunogen Recombinant fragment within Human NAPSA aa 1-150.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9476
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.