Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Napsin A Antibody (NAPSA/1239), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2452450.2MG
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Napsin A Monoclonal antibody specifically detects Napsin A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, SDS-Page, Immunofluorescence.Specifications
Napsin A | |
Monoclonal | |
0.2mg/mL | |
Western Blot 1 - 2ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml | |
Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02 | |
Mouse | |
37 kDa | |
0.2 mg | |
Cancer, Cell Biology, Cellular Markers | |
9476 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
NAPSA/1239 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
NAPSA | |
Recombinant fragment of human Napsin-A protein | |
Protein A purified | |
RUO | |
Primary | |
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B. There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs. | |
Store at 4C. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction