Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Napsin A Rabbit anti-Human, Clone: NAPSA/4400R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30786920UG
Description
Napsin A Monoclonal specifically detects Napsin A in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Napsin A | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02 | |
Recombinant fragment (aa50-150) of human Napsin A protein (exact sequence is proprietary) | |
20 μg | |
Cancer, Cell Biology, Cellular Markers | |
9476 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin), Peptide Array | |
NAPSA/4400R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction