Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Naratuximab Chimeric Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542100 Supplier Invitrogen™ Supplier No. MA542100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Naratuximab is a biosimilar that targets CD37. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Naratuximab Chimeric
Applications ELISA
Classification Recombinant Monoclonal
Concentration 3.19 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias K7153A
Host Species Human
Immunogen Human CD37/TSPAN26.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.