Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARC-1 (PCSK9) Mouse anti-Human, Unconjugated, Clone: 15A6, MilliporeSigma™

Catalog No. MABS21611MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS21611MI 100 μg
MABS21612MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS21611MI Supplier MilliporeSigma Supplier No. MABS2161100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NARC-1 (PCSK9) Monoclonal antibody specifically detects NARC-1 (PCSK9) in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARC-1 (PCSK9)
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 15A6
Conjugate Unconjugated
Gene Accession No. NP_777596
Gene Alias Proprotein convertase subtilisin/kexin type 9;Neural apoptosis-regulated convertase 1;Proprotein convertase 9;PC9;Subtilisin/kexin-like protease PC9.
Gene Symbols PCSK9, NARC1, PSEC0052
Host Species Mouse
Immunogen Full-length purified human Proprotein convertase 9 (PCSK9).
Molecular Weight of Antigen ∼80 and 60 kDa observed (proprotein and cleaved form); 74.29 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 15A6 is a mouse monoclonal antibody that specifically detects Proprotein convertase 9 (NARC-1) and its cleaved form in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.