Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nardilysin (NRDC) Mouse anti-Human, Unconjugated, Clone: 102, MilliporeSigma™

Catalog No. MABS20562MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS20562MI 25 μL
MABS20561MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS20562MI Supplier MilliporeSigma Supplier No. MABS205625UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nardilysin (NRDC) Monoclonal antibody specifically detects Nardilysin (NRDC) in Human samples. It is validated for Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nardilysin (NRDC)
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 102
Conjugate Unconjugated
Gene Accession No. NP_002516.2
Gene Alias N-arginine dibasic convertase;NRD convertase;NRDC;NRD-C;Nardilysin convertase
Gene Symbols NRDC, NRD1
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 80 amino acids from the N-terminal of human Nardilysin.
Molecular Weight of Antigen ∼140 kDa observed; 131.70 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 102 is a mouse monoclonal antibody that specifically detects human Nardilysin. It targets an epitope within 80 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.