Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nardilysin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530213 Supplier Invitrogen™ Supplier No. PA530213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, HeLa, mouse kidney. Predicted reactivity: Rat (97%), Chicken (86%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cleaves peptide substrates on the N-terminus of arginine residues in dibasic pairs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nardilysin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Nrdc
Gene Accession No. O43847, Q8BHG1
Gene Alias 2600011I06Rik; AI875733; hNRD1; hNRD2; nardilysin; nardilysin (N-arginine dibasic convertase); nardilysin 1; nardilysin 1 (N-arginine dibasic convertase); nardilysin convertase; nardilysin, N-arginine dibasic convertase 1; nardilysin, N-arginine dibasic convertase, NRD convertase 1; N-arginine dibasic convertase; N-arginine dibasic convertase 1; NRD convertase; Nrd1; NRDC; NRD-C
Gene Symbols Nrd1, Nrdc
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human Nardilysin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230598, 4898
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.