Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nardilysin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598016
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598016 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598016 Supplier Invitrogen™ Supplier No. PA598016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nardilysin Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Cleaves peptide substrates on the N-terminus of arginine residues in dibasic pairs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nardilysin
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Nrdc
Gene Accession No. O43847
Gene Alias 2600011I06Rik; AI875733; hNRD1; hNRD2; nardilysin; nardilysin (N-arginine dibasic convertase); nardilysin 1; nardilysin 1 (N-arginine dibasic convertase); nardilysin convertase; nardilysin, N-arginine dibasic convertase 1; nardilysin, N-arginine dibasic convertase, NRD convertase 1; N-arginine dibasic convertase; N-arginine dibasic convertase 1; NRD convertase; Nrd1; NRDC; NRD-C
Gene Symbols Nrdc
Host Species Rabbit
Immunogen Recombinant Human Nardilysin protein (32-113AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4898
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.