Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NARF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565754 Supplier Invitrogen™ Supplier No. PA565754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTDDQENVSAD APSPAQENGE KGEFHKLADA KIFLSDCLAC DSCMTAEEGV QLSQQNAKDF FRVLNLNKKC DTSKHKVLVV SVCPQSLPYF AAKFNLSVTD ASRRLCGFLK SLG Highest antigen sequence identity to the following orthologs - mouse 84%, rat 93%.

Several proteins have been found to be prenylated and methylated at their carboxyl-terminal ends. Prenylation was initially believed to be important only for membrane attachment. However, another role for prenylation appears to be its importance in protein-protein interactions. The only nuclear proteins known to be prenylated in mammalian cells are prelamin A- and B-type lamins. Prelamin A is farnesylated and carboxymethylated on the cysteine residue of a carboxyl-terminal CaaX motif. This post-translationally modified cysteine residue is removed from prelamin A when it is endoproteolytically processed into mature lamin A. The protein encoded by this gene binds to the prenylated prelamin A carboxyl-terminal tail domain. It may be a component of a prelamin A endoprotease complex. The encoded protein is located in the nucleus, where it partially colocalizes with the nuclear lamina. It shares limited sequence similarity with iron-only bacterial hydrogenases. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene, including one with a novel exon that is generated by RNA editing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NARF
Gene Accession No. Q9UHQ1
Gene Alias 4430402O11Rik; IOP2; iron-only hydrogenase-like protein 2; Narf; Nuclear prelamin A recognition factor; prenyl-dependent prelamin A binding protein; RGD1310894
Gene Symbols NARF
Host Species Rabbit
Immunogen Recombinant Human NARF. Recombinant protein control fragment (Product #RP-104958).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26502
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.