Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NARFL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559325 Supplier Invitrogen™ Supplier No. PA559325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLEASRPDFF NQEHQTRDVD CVLTTGEVFR LLEEEGVSLP DLEPAPLDSL CSGASAEEPT SHRGGGSGGY LEHVFRHAAR ELFGIHVAEV TYKP Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into extramitochondrial Fe/S proteins. Seems to negatively regulate the level of HIF1A expression, although this effect could be indirect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARFL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Narfl
Gene Accession No. Q9H6Q4
Gene Alias 9030612I22Rik; AI504405; CIAO3; Cytosolic Fe-S cluster assembly factor NARFL; Cytosolic iron-sulfur assembly component 3; HPRN; IOP1; iron-only hydrogenase-like protein 1; LET1 like/JFP15; LET1L; NAR1; NARFL; nuclear prelamin A recognition factor like; nuclear prelamin A recognition factor-like; nuclear prelamin A recognition factor-like protein; PRN; protein related to Narf
Gene Symbols Narfl
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NARFL. Recombinant protein control fragment (Product #RP-98667).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64428
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.