Learn More
NARFL Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305770AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Orangutan
Component of the cytosolic iron-sulfur (Fe/S) protein assembly machinery. Required for maturation of extramitochondrial Fe/S proteins. Seems to negatively regulate the level of HIF1A expression, although this effect could be indirect.Specifications
NARFL | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9H6Q4 | |
NARFL | |
between 225 and 275 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
NARFL | |
HPRN, IOP1, LET1L, PRN, LET1 like/JFP15, cytosolic Fe-S cluster assembly factor NARFL, iron-only hydrogenase-like protein 1, nuclear prelamin A recognition factor-like protein, protein related to Narf | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64428 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.