Learn More
NARG1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | NARG1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2414
![]() |
Bethyl Laboratories, Inc
IHC00618T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2413
![]() |
Bethyl Laboratories, Inc
IHC00618 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Crab-eating macaque
This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.Specifications
NARG1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Ga19, NARG1, NATH, TBDN100, N-alpha-acetyltransferase 15, NatA auxiliary subunit, N-terminal acetyltransferase, NMDA receptor regulated 1, NMDA receptor-regulated protein 1, gastric cancer antigen Ga19, protein tubedown-1, transcriptional coactivator tubedown-100, tubedown-1 | |
NAA15 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9BXJ9 | |
80155 | |
Between 816 and 866 | |
Primary | |
4° C | |
NAA15 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.