Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARG1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984276
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4276 Supplier Bioss Supplier No. BS19020R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.

Specifications

Antigen NARG1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NAA15
Gene Accession No. Q9BXJ9
Gene Alias 5730450D16Rik; 6330400I15; ASTBDN; GA19; Gastric cancer antigen Ga19; mNAT1; N(alpha)-acetyltransferase 15, NatA auxiliary subunit; NAA15; N-alpha-acetyltransferase 15, NatA auxiliary subunit; NARG1; NAT1P; NATH; NMDA receptor regulated 1; NMDA receptor-regulated gene 1; NMDA receptor-regulated protein 1; N-terminal acetyltransferase; N-terminal acetyltransferase 1; N-terminal aceyltransferase 1; Protein tubedown-1; TBDN; Tbdn-1; TBDN100; transcriptional coactivator tubedown-100; tubedown; tubedown-1
Gene Symbols NAA15
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NARG1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80155
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.