Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARG1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734140 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4140 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4140 Supplier Proteintech Group Inc Supplier No. 238451AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

N-alpha-acetyltransferase 15, NatA auxiliary subunit also known as gastric cancer antigen Ga19 (GA19), NMDA receptor-regulated protein 1 (NARG1), and Tbdn100 is a protein that in humans is encoded by the NAA15 gene.NARG1 is the auxiliary subunit of the NatA (Nalpha-acetyltransferase A) complex. This NatA complex can associate with the ribosome and catalyzes the transfer of an acetyl group to the Nalpha-terminal amino group of proteins as they emerge from the exit tunnel.

Specifications

Antigen NARG1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NAA15
Gene Accession No. Q9BXJ9
Gene Alias Ga19, Gastric cancer antigen Ga19, N terminal acetyltransferase, NAA15, NARG1, NATH, Protein tubedown 1, TBDN100
Gene Symbols NAA15
Host Species Rabbit
Immunogen NARG1 Fusion Protein Ag20842
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80155
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.