Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NARG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524021
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524021 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524021 Supplier Invitrogen™ Supplier No. PA524021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NARG2 Polyclonal Antibody for Western Blot

NARG2 is expressed at relatively high levels in dividing and immature cells, and is down-regulated upon terminal differentiation. NARG2 is a novel (S/T)PXX motif-containing nuclear protein; this motif is present in many transcription factors as well as other regulatory proteins that bind to DNA such as histones and RNA polymerase II. Three different isoforms exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARG2
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ICE2
Gene Accession No. Q3UZ18, Q659A1
Gene Alias B230343B06Rik; BRCC1; breast cancer cell 1; Ice2; interactor of little elongation complex ELL subunit 2; interactor of little elongator complex ELL subunit 2; Little elongation complex subunit 2; NARG2; NMDA receptor regulated 2; NMDA receptor-regulated gene 2; NMDA receptor-regulated protein 2; UNQ3101/PRO10100
Gene Symbols ICE2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 26-54 amino acids from the N-terminal region of human NARG2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79664, 93697
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.