Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniStore™ Reagent Bottles, Narrow Mouth, PP, Bulk Packed
SDPSCI_ED

Catalog No. S43673 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S43673 250 mL
S43669 15 mL
S43670 30 mL
S43671 60 mL
S43672 125 mL
S43674 500 mL
S43675 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S43673 Supplier United Scientific™ Supplier No. 33303BULK
Add to Cart
Add to Cart

Narrow Mouth Polypropylene Reagent Bottles are rigid, translucent and have fine contact clarity.

UniStore™ Narrow Mouth, PP, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.

  • Excellent chemical resistance to most acids, bases, and alcohols.
  • Polypropylene caps are included.
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
  • Leakproof closure with integrated seal ring.
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces.
  • Certificates of Analysis for leakproof test details are available upon request.

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Translucent
Diameter (Metric) Inner Neck 18 mm
Material Polypropylene
Packaging Bulk
Quantity 250 Pk.
Sterility Non-sterile
Product Line UniStore
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 133 mm
Product Type Reagent Bottle
Height (English) 5.23 in.
Closure Material Polypropylene
Closure Size 24 mm
Diameter (Metric) 18 mm
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.