Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Reagent Bottles
SCI_ED

Catalog No. S24488
Click to view available options
Mouth:
Narrow
Wide

Rigid bottles for universities and industrial labs

  • Made from polypropylene
  • Available as wide or narrow neck
  • Autoclavable
  • Leak-proof

Specifications

Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Clear
Description Leak-proof
Material Polypropylene
Length (Metric) 6.98 cm
Quantity 1 Ea.
Sterility Non-sterile
Width (Metric) 6.98 cm
Closure Included Yes
Autoclavable Autoclavable
Graduated No
Mouth Narrow
Height (Metric) 17.14 cm
Product Type Reagent Bottle
Height (English) 6.75 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.