Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NARS Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | NARS |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118966
|
Invitrogen™
PA5118966 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121748
|
Invitrogen™
PA5121748 |
20 μL |
Each for $209.50
|
|
|||||
Description
NARS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Asparaginyl-tRNA synthetase is localized to the cytoplasm and belongs to the class II family of tRNA synthetases. The N-terminal domain represents the signature sequence for the eukaryotic asparaginyl-tRNA synthetases.Specifications
NARS | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
NARS1 | |
3010001M15Rik; AA960128; AsnRS; asparagine tRNA ligase 1, cytoplasmic; asparagine--tRNA ligase, cytoplasmic; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase, cytoplasmic; C78150; LRRGT00113; Nars; NARS1 | |
NARS1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O43776 | |
4677 | |
Recombinant protein Asparaginyl tRNA Synthetase. The antigen corresponds to amino acid range 1-330 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title