Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114179 Supplier Invitrogen™ Supplier No. PA5114179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TIYVDSQKEN ERWNVISKSQ LKNIKKMWHR EQMKSESREK KEAEDSLRRE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 86%; Rat: 92%; Zebrafish: 75%.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Asparaginyl-tRNA synthetase is localized to the cytoplasm and belongs to the class II family of tRNA synthetases. The N-terminal domain represents the signature sequence for the eukaryotic asparaginyl-tRNA synthetases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NARS1
Gene Accession No. O43776
Gene Alias 3010001M15Rik; AA960128; AsnRS; asparagine tRNA ligase 1, cytoplasmic; asparagine--tRNA ligase, cytoplasmic; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase, cytoplasmic; C78150; LRRGT00113; Nars; NARS1
Gene Symbols NARS1
Host Species Rabbit
Immunogen Immunogen sequence: TIYVDSQKEN ERWNVISKSQ LKNIKKMWHR EQMKSESREK KEAEDSLRRE (aa 43-92).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4677
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.