Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116533 Supplier Invitrogen™ Supplier No. PA5116533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-29, 293T, LO2, Jurkat, Mouse pancreas Immunogen sequence: PALGDVFCIA QSYRAEQSRT RRHLAEYTHV EAECPFLTFD DLLNRLEDLV CDVVDRILKS PAGSIVHELN PNFQPPKRPF KRMNYSDAIV WLKEHDVKKE DGTFYEFGED IPEAPERLMT DTINEPILLC RFPVEIKSFY MQRCPEDSRL TESVDVLMPN VGEIVGGSMR IFDSEEILAG YKREGIDPTP YYWYTDQRKY GTCPHGGYGL GLERFLTWIL NRYHIRDVCL YPRFVQRCTP.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Asparaginyl-tRNA synthetase is localized to the cytoplasm and belongs to the class II family of tRNA synthetases. The N-terminal domain represents the signature sequence for the eukaryotic asparaginyl-tRNA synthetases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NARS1
Gene Accession No. O43776, Q8BP47
Gene Alias 3010001M15Rik; AA960128; AsnRS; asparagine tRNA ligase 1, cytoplasmic; asparagine--tRNA ligase, cytoplasmic; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase, cytoplasmic; C78150; LRRGT00113; Nars; NARS1
Gene Symbols NARS1
Host Species Rabbit
Immunogen Recombinant protein (or fragment)).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4677, 70223
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.