Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NARS2 Antibody (OTI10C10), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272876AF350
Description
NARS2 Monoclonal specifically detects NARS2 in Human samples. It is validated for Western Blot.Specifications
NARS2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
NARS2 | |
Full length recombinant protein of human NARS2 (NP_078954) produced in HEK293T cells. | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI10C10 | |
Western Blot | |
asnRS, asparagine tRNA ligase 2, mitochondrial (putative), Asparagine--tRNA ligase, asparaginyl-tRNA synthetase 2, mitochondrial (putative), EC 6.1.1, EC 6.1.1.22, FLJ23441, probable asparaginyl-tRNA synthetase, mitochondrial, SLM5 | |
Mouse | |
0.1 mL | |
79731 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction