Learn More
Invitrogen™ NARS2 Monoclonal Antibody (OTI5A9)

Description
NARS2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | NARS2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI5A9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NARS2 |
| Gene Accession No. | Q96I59 |
| Gene Alias | AI875199; asnRS; asparagine tRNA ligase 2, mitochondrial (putative); asparagine--tRNA ligase; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase 2 (mitochondrial)(putative); asparaginyl-tRNA synthetase 2, mitochondrial; asparaginyl-tRNA synthetase 2, mitochondrial (putative); deafness, autosomal recessive 94; DFNB94; EC 6.1.1.22; Nars2; Nars2, similar to CG6796-PA; probable asparagine--tRNA ligase, mitochondrial; probable asparaginyl-tRNA synthetase, mitochondrial; RGD1305145; SLM5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.