Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555420 Supplier Invitrogen™ Supplier No. PA555420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAEFYMIEAE ISFVDSLQDL MQVIEELFKA TTMMVLSKCP EDVELCHKFI APGQKDRLEH MLKNNFLIIS YTEAVEILKQ ASQNFTFTPE WGADLRTEHE KYLVKHCGNI PVFVINYPLT LKPFYMRDNE DGPQHTVAAV DLLVPGVG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 78%.

Asparagine-tRNA ligase (EC 6.1.1.22) catalyzes the chemical reaction: ATP + L-asparagine + tRNAAsn <-> AMP + diphosphate + L-asparaginyl-tRNAAsn. This enzyme participates in alanine and aspartate metabolism and aminoacyl-tRNA biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NARS2
Gene Accession No. Q96I59
Gene Alias AI875199; asnRS; asparagine tRNA ligase 2, mitochondrial (putative); asparagine--tRNA ligase; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase 2 (mitochondrial)(putative); asparaginyl-tRNA synthetase 2, mitochondrial; asparaginyl-tRNA synthetase 2, mitochondrial (putative); deafness, autosomal recessive 94; DFNB94; EC 6.1.1.22; Nars2; Nars2, similar to CG6796-PA; probable asparagine--tRNA ligase, mitochondrial; probable asparaginyl-tRNA synthetase, mitochondrial; RGD1305145; SLM5
Gene Symbols NARS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NARS2. Recombinant protein control fragment (Product #RP-101021).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.