Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NARX Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144475 Supplier Invitrogen™ Supplier No. PA5144475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NARX Polyclonal Antibody for Western Blot, ELISA

Acts as a sensor for nitrate/nitrite and transduces signal of nitrate availability to the NarL protein and of both nitrate/nitrite to the NarP protein. NarX probably activates NarL and NarP by phosphorylation in the presence of nitrate. NarX also plays a negative role in controlling NarL activity, probably through dephosphorylation in the absence of nitrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARX
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene narX
Gene Accession No. P0AFA2
Gene Alias narR
Gene Symbols narX
Host Species Rabbit
Immunogen Recombinant Escherichia coli (strain K12) NARX protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75203337
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.