Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NASP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593299 Supplier Invitrogen™ Supplier No. PA593299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WDMLDLAKII FKRQETKEAQ LYAAQAHLKL GEVSVESENY VQAVEEFQSC LNLQEQYLEA HDRLLAETHY QLGLAYGYNS QYDEAVAQFS KSIEVIENRM AVLNEQVKEA EGSSAEYKKE IEELKELLPE IREKIEDAKE SQRSGNVAEL ALKATLVESS TSGFTPGGGG SSVSMIASRK PTDGASSSNC VTDISHLVRK KRKPEEESPR KDDAKKAKQE PEVNGGSGDA VPSGNEVSEN MEEEAENQAE SRAAVEGTVE AGATVESTAC; Positive Samples: SW620, HL-60, 293T; Cellular Location: Cytoplasm, Nucleus.

NASP encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NASP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NASP
Gene Accession No. P49321, Q66HD3, Q99MD9
Gene Alias 5033430J04Rik; AI131596; AI317140; D4Ertd767e; DKFZp547F162; EMBL:AAH81913.1}; Epcs32; FLB7527; FLJ31599; FLJ35510; histone H1-binding protein; HMDRA1; MGC19722; MGC20372; MGC2297; Nasp; nasp {ECO:0000312; NASP histone chaperone; nasp protein; Nasp-T; Nuclear autoantigenic sperm protein; nuclear autoantigenic sperm protein (histone-binding); PRO1999; somatic histone binding protein NASP; wu:fd20g12; zgc:56007; zgc:85651
Gene Symbols NASP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-449 of human NASP (NP_6895112).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298441, 4678, 50927
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.