Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NASP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118100 Supplier Invitrogen™ Supplier No. PA5118100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NASP Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, IP

NASP encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NASP
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene NASP
Gene Accession No. P49321
Gene Alias 5033430J04Rik; AI131596; AI317140; D4Ertd767e; DKFZp547F162; EMBL:AAH81913.1}; Epcs32; FLB7527; FLJ31599; FLJ35510; histone H1-binding protein; HMDRA1; MGC19722; MGC20372; MGC2297; Nasp; nasp {ECO:0000312; NASP histone chaperone; nasp protein; Nasp-T; Nuclear autoantigenic sperm protein; nuclear autoantigenic sperm protein (histone-binding); PRO1999; somatic histone binding protein NASP; wu:fd20g12; zgc:56007; zgc:85651
Gene Symbols NASP
Host Species Rabbit
Immunogen E. coli-derived Human NASP fragment.
Purification Method Protein A, Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4678
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.