Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NASP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556494 Supplier Invitrogen™ Supplier No. PA556494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTDVQEECRE KGGQEKQGEV IVSIEEKPKE VSEEQPVVTL EKQGTAVEVE AESLDPTVKP VDVGGDEPEE KVVTSENEAG KAVLEQLVGQ EVPPAE Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 48%.

NASP encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NASP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NASP
Gene Accession No. P49321
Gene Alias 5033430J04Rik; AI131596; AI317140; D4Ertd767e; DKFZp547F162; EMBL:AAH81913.1}; Epcs32; FLB7527; FLJ31599; FLJ35510; histone H1-binding protein; HMDRA1; MGC19722; MGC20372; MGC2297; Nasp; nasp {ECO:0000312; NASP histone chaperone; nasp protein; Nasp-T; Nuclear autoantigenic sperm protein; nuclear autoantigenic sperm protein (histone-binding); PRO1999; somatic histone binding protein NASP; wu:fd20g12; zgc:56007; zgc:85651
Gene Symbols NASP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NASP. Recombinant protein control fragment (Product #RP-94205).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4678
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.