Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560474 Supplier Invitrogen™ Supplier No. PA560474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRNKLDLETL TDILQHQIRA VPFENLN Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 89%.

Arylamine N-acetyltransferases (NAT-1 and NAT-2) catalyze N- or O-acetylation of heterocyclic and arylamine substrates in the detoxification of a wide array of drugs. Certain alleles causing high N-acetyltransferase activity have been associated with colon and urinary bladder cancers, as NATs also bioactivate several known carcinogens. Both NAT-1 and NAT-2 are cytoplasmic proteins and play an active role in the detoxification of many arylamine and hydrazine drugs. N-acetylation polymorphism is determined by the level of NAT activity in liver tissues, and has been linked to the action and toxicity of drugs that contain amines. Human NAT-1 is the functional homolog of rodent NAT-2, while human NAT-2 is the functional homolog of rodent NAT-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAT1
Gene Accession No. P18440
Gene Alias AAC1; Arylamide acetylase 1; arylamine N-acetyltransferase 1; AT-1; MNAT; Monomorphic arylamine N-acetyltransferase; N(alpha)-acetyltransferase 15, NatA auxiliary subunit; N-acetyl transferase 1; N-acetyl transferase 1, liver, blood; N-acetyltransferase (arylamine N-acetyltransferase); N-acetyltransferase 1; N-acetyltransferase 1 (arylamine N-acetyltransferase); N-acetyltransferase type 1; Nat; Nat1; NAT-1; NATI
Gene Symbols NAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NAT1. Recombinant protein control fragment (Product #RP-99582).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.