Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT10 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB268739 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB268739 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB268739 Supplier Novus Biologicals Supplier No. NBP232263F
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NAT10 Polyclonal antibody specifically detects NAT10 in Human, Mouse samples. It is validated for Immunohistochemistry

Specifications

Antigen NAT10
Applications Immunohistochemistry
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS
Gene Alias ALP, DKFZp434C116, EC 2.3.1.-, FLJ10774, FLJ12179, FLJ23850, hALP, KIAA1709, N-acetyltransferase 10, N-acetyltransferase 10 (GCN5-related), N-acetyltransferase-like protein, NET43
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 650 to 700 of human N-acetyltransferase 10 using the numbering given in entry NP_078938.2 (GeneID 55226).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Direct Reversal of DNA Damage, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55226
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.