Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT10 Antibody (SR1073), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322400100UL
Description
NAT10 Monoclonal antibody specifically detects NAT10 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
NAT10 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1073 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
ALP, DKFZp434C116, EC 2.3.1.-, FLJ10774, FLJ12179, FLJ23850, hALP, KIAA1709, N-acetyltransferase 10, N-acetyltransferase 10 (GCN5-related), N-acetyltransferase-like protein, NET43 | |
A synthesized peptide derived from human NAT10 (Uniprot #: Q9H0A0) | |
100 μg | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Direct Reversal of DNA Damage, Immunology, Signal Transduction | |
55226 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction