Learn More
Invitrogen™ NAT10 Recombinant Rabbit Monoclonal Antibody (5Z3E5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA537909
Description
Positive Samples: HeLa, K-562, SH-SY5Y, Mouse testis Immunogen sequence: KTLTDEDEAD QGGWLAAFWK DFRRRFLALL SYQFSTFSPS LALNIIQNRN MGKPAQPALS REELEALFLP YDLKRLEMYS RNMVDYHLIM DMIPAISRIY FLNQLGDLAL SAAQSALLLG IGLQHKSVDQ LEKEIELPSG QLMGLFNRII RKVVKLFNEV QEKAIEEQMV AAKDVVMEPT MKTLSDDLDE AAKEFQEKHK KEVGKLKSMD LSEYIIRGDD EEWNEVLNKA GPNASIISLK SDKKRKLEAK QEPKQSKKLK NRETKNKKDM KLKRKK.
IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.
Specifications
NAT10 | |
Recombinant Monoclonal | |
0.6 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K224, Q9H0A0 | |
Nat10 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 750-1025 of human NAT10 (Q9H0A0). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5Z3E5 | |
Unconjugated | |
Nat10 | |
18S rRNA cytosine acetyltransferase; AI429152; ALP; hALP; KIAA1709; N-acetyltransferase 10; N-acetyltransferase 10 (GCN5-related); N-acetyltransferase-like protein; NAT10; NET43; novel putative ATPase domain containing protein; RGD1306717; RNA cytidine acetyltransferase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
311257, 55226, 98956 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.