Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAT13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592334
Description
Immunogen sequence: MKGSRIELGD VTPHNIKQLK RLNQVIFPVS YNDKFYKDVL EVGELAKLAY FNDIAVGAVC CRVDHSQNQK RLYIMTLGCL APYRRLGIGT KMLNHVLNIC EKDGTFDNIY LHVQISNESA IDFYRKFGFE IIETKKNYYK RIEPADAHVL QKNLKVPSGQ NADVQKTDN; Positive Samples: SKOV3, HL-60, 293T, MCF7; Cellular Location: Cytoplasm.
NAT13 is a probable catalytic component of the ARD1A-NARG1 complex which displays alpha (N-terminal) acetyltransferase activity.
Spécifications
NAT13 | |
Polyclonal | |
Unconjugated | |
NAA50 | |
2600005K24Rik; 2810441M03Rik; AW112078; hNaa50p; hNAT5; hSAN; MAK3; Mak3 homolog; Mak3p; Mak3p homolog; N(alpha)-acetyltransferase 50, NatE catalytic subunit; Naa50; N-acetyltransferase 13; N-acetyltransferase 13 (GCN5-related); N-acetyltransferase 5; N-acetyltransferase NAT13; N-acetyltransferase san homolog; Nalpha acetyltransferase 50; N-alpha-acetyltransferase 50; N-alpha-acetyltransferase 50, NatE catalytic subunit; Nat13; NAT13P; Nat5; NAT5P; NatE catalytic subunit; N-epsilon-acetyltransferase 50; San | |
Rabbit | |
Affinity chromatography | |
RUO | |
288108, 72117, 80218 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6PGB6, Q9GZZ1 | |
NAA50 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-169 of human NAA50 (NP_0794221). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Sécurité et manipulation
WARNING: Cancer - www.P65Warnings.ca.gov
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'
missing translation for 'spotOppurtunityImprovement'missing translation for 'shareContentCorrection'