Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596661 Supplier Invitrogen™ Supplier No. PA596661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKGSRIELGD VTPHNIKQLK RLNQVIFPVS YNDKFYKDVL EVGELAKLAY FNDIAVGAVC CRVDHSQNQK RLYIMTLGCL APYRRLGIGT KMLNHVLNIC EKDGTFDNIY LHVQISNESA IDFYRKFGFE IIETKKNYYK RIEPADAHVL QKNLKVPSGQ NADVQKTDN; Positive Samples: SKOV3, HL-60, 293T, MCF7, SW480, Mouse brain; Cellular Location: Cytoplasm.

NAT13 is a probable catalytic component of the ARD1A-NARG1 complex which displays alpha (N-terminal) acetyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT13
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NAA50
Gene Accession No. Q6PGB6, Q9GZZ1
Gene Alias 2600005K24Rik; 2810441M03Rik; AW112078; hNaa50p; hNAT5; hSAN; MAK3; Mak3 homolog; Mak3p; Mak3p homolog; N(alpha)-acetyltransferase 50, NatE catalytic subunit; Naa50; N-acetyltransferase 13; N-acetyltransferase 13 (GCN5-related); N-acetyltransferase 5; N-acetyltransferase NAT13; N-acetyltransferase san homolog; Nalpha acetyltransferase 50; N-alpha-acetyltransferase 50; N-alpha-acetyltransferase 50, NatE catalytic subunit; Nat13; NAT13P; Nat5; NAT5P; NatE catalytic subunit; N-epsilon-acetyltransferase 50; San
Gene Symbols NAA50
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-169 of human NAA50 (NP_0794221).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288108, 72117, 80218
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.