Learn More
Invitrogen™ NAT13 Polyclonal Antibody

Description
Positive Control: A431 cell lysate, Hela cell lysate, mouse stomach tissue lysate, rat cerebellum tissue lysate, rat brain tissue, human thyroid tissue, rat cerebellum tissue, human skin tissue, human breast carcinoma tissue, Daudi. Subcellular Location: Cytoplasm, Nucleus.
Specifications
Specifications
| Antigen | NAT13 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | NAA50 |
| Gene Accession No. | Q6PGB6, Q9GZZ1 |
| Gene Alias | 2600005K24Rik; 2810441M03Rik; AW112078; hNaa50p; hNAT5; hSAN; MAK3; Mak3 homolog; Mak3p; Mak3p homolog; N(alpha)-acetyltransferase 50, NatE catalytic subunit; Naa50; N-acetyltransferase 13; N-acetyltransferase 13 (GCN5-related); N-acetyltransferase 5; N-acetyltransferase NAT13; N-acetyltransferase san homolog; Nalpha acetyltransferase 50; N-alpha-acetyltransferase 50; N-alpha-acetyltransferase 50, NatE catalytic subunit; Nat13; NAT13P; Nat5; NAT5P; NatE catalytic subunit; N-epsilon-acetyltransferase 50; San |
| Gene Symbols | NAA50 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.