Learn More
Invitrogen™ NAT14 Polyclonal Antibody

Description
Immunogen sequence: PSHLSVREMRE DEKPLVLEML KAGVKDTENR VALHALTRPP ALLLLAAASS GL Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
Specifications
Specifications
| Antigen | NAT14 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NAT14 |
| Gene Accession No. | Q8WUY8 |
| Gene Alias | BC047219; K562 cell-derived leucine-zipper-like protein 1; K562 cells-derived leucine zipper-like protein 1; K562 cells-derived leucine-zipper-like protein 1; KLP1; N-acetyltransferase 14; N-acetyltransferase 14 (GCN5-related, putative); N-acetyltransferase 14 (putative); Nat14; RGD1565991; Zfp628; zinc finger protein 628 |
| Gene Symbols | NAT14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.