Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115617 Supplier Invitrogen™ Supplier No. PA5115617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NAT14.

Probable acetyltransferase that binds the 5'-GGACTACAG-3' sequence of coproporphyrinogen oxidase promoter. Able to activate transcription of a reporter construct in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NAT14
Gene Accession No. Q8BVG8, Q8WUY8
Gene Alias BC047219; K562 cell-derived leucine-zipper-like protein 1; K562 cells-derived leucine zipper-like protein 1; K562 cells-derived leucine-zipper-like protein 1; KLP1; N-acetyltransferase 14; N-acetyltransferase 14 (GCN5-related, putative); N-acetyltransferase 14 (putative); Nat14; RGD1565991; Zfp628; zinc finger protein 628
Gene Symbols NAT14
Host Species Rabbit
Immunogen A synthesized peptide derived from human NAT14(Accession Q8WUY8), corresponding to amino acid residues A163-L206.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269854, 361500, 57106
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.