Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT16 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549546
Description
Probable N-acetyltransferase. Shows only trace activity toward L-His and no N-acetyltransferase activity toward other amino acids. The physiological substrate of this enzyme is unknown.
Specifications
NAT16 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8N8M0 | |
NAT16 | |
KLH conjugated synthetic peptide between 12-46 amino acids from human NAT16. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NAT16 | |
Putative N-acetyltransferase 16, 2.3.1.-, NAT16, C7orf52 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375607 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction