Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT2 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5145440
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145440 20 μL
FEPA5145439 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145440 Supplier Invitrogen™ Supplier No. PA5145440
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Participates in the detoxification of a plethora of hydrazine and arylamine drugs. Catalyzes the N- or O-acetylation of various arylamine and heterocyclic amine substrates and is able to bioactivate several known carcinogens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene NAT2
Gene Accession No. P11245, P50295, P50298
Gene Alias Aac2; Arylamide acetylase 2; arylamine N-acetyltransferase 2; AT-2; AV377607; N-acetyl transferase 2; N-acetyltransferase 2; N-acetyltransferase 2 (arylamine N-acetyltransferase); N-acetyltransferase type 2; N-Acetyltransferase-2 (arylamine N-acetyltransferase); Nat2; NAT-2; Nat2a; PNAT; Polymorphic arylamine N-acetyltransferase
Gene Symbols NAT2
Host Species Rabbit
Immunogen Recombinant Rat NAT2 protein, Asp20-Val280 (Accession #P50298).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10, 116632, 17961
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.