Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT8 Monoclonal Antibody (OTI5A8)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524951

Catalog No. PIMA524951


Only null left
Add to Cart

Description

Description

NAT8 Monoclonal Antibody for ICC/IF, IHC (P)

This protein, isolated using the differential display method to detect tissue-specific genes, is specifically expressed in kidney and liver. The encoded protein shows amino acid sequence similarity to N-acetyltransferases. A similar protein in Xenopus affects cell adhesion and gastrulation movements, and may be localized in the secretory pathway. A highly similar paralog is found in a cluster with this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NAT8
Monoclonal
1.00 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHE5
NAT8
Full-length protein expressed in 293T cell transfected with human NAT8 expression vector.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI5A8
Unconjugated
NAT8
0610037O16Rik; Acetyltransferase 2; ATase2; camello-like 4; camello-like protein 1; Camello-like protein 4; CCNAT; CML1; Cml4; Cysteinyl-conjugate N-acetyltransferase; GLA; Hcml1; kidney- and liver-specific gene product; N-acetyltransferase 8; N-acetyltransferase 8 (camello like); N-acetyltransferase 8 (GCN5-related, putative); N-acetyltransferase 8 (putative); NAT8; probable N-acetyltransferase 8; probable N-acetyltransferase CML4; putative N-acetyltransferase Camello 4; TSC501; TSC510
Mouse
Affinity Chromatography
RUO
9027
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.