Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596792 Supplier Invitrogen™ Supplier No. PA596792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, Mouse spleen, Mouse testis, Mouse lung, Rat kidney; Cellular Location: Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Single-pass type II membrane protein Immunogen sequence: LLALVFSISL FPALWFLAKK PWTEYVDMTL CTDMSDITKS YLSERGSCFW VAESEEKVVG MVGALPVDDP TLREKRLQLF HLFVDSEHRR QGIAKALVRT VLQFARDQGY SEVILDTGTI QLSAMALYQS MGFKKTGQSF FCVWARLVAL HTVHFIYHLP.

This protein, isolated using the differential display method to detect tissue-specific genes, is specifically expressed in kidney and liver. The encoded protein shows amino acid sequence similarity to N-acetyltransferases. A similar protein in Xenopus affects cell adhesion and gastrulation movements, and may be localized in the secretory pathway. A highly similar paralog is found in a cluster with this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT8
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NAT8
Gene Accession No. Q9JIY7, Q9QXT3, Q9UHE5
Gene Alias 0610037O16Rik; Acetyltransferase 2; ATase2; camello-like 4; camello-like protein 1; Camello-like protein 4; CCNAT; CML1; Cml4; Cysteinyl-conjugate N-acetyltransferase; GLA; Hcml1; kidney- and liver-specific gene product; N-acetyltransferase 8; N-acetyltransferase 8 (camello like); N-acetyltransferase 8 (GCN5-related, putative); N-acetyltransferase 8 (putative); NAT8; probable N-acetyltransferase 8; probable N-acetyltransferase CML4; putative N-acetyltransferase Camello 4; TSC501; TSC510
Gene Symbols NAT8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 61-220 of human NAT8 (Q9UHE5).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64570, 68396, 9027
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.