Learn More
Invitrogen™ NAT8B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596607
Description
Immunogen sequence: WFLAKKPWTR YVDIALRTDM SDITKSYLSE CGSCFWVGES EEKVVGTVGA LPVDDPTLRE KRLQLFHLSV DNEHRGQGIA KALVRTVLQF A; Positive Samples: Rat brain; Cellular Location: Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Single-pass type II membrane protein.
The protein encoded by this gene is highly similar to the N-acetyltransferase 8 (NAT8) gene product, which is a kidney and liver protein with homology to bacterial acetyltransferases involved in drug resistance. This gene is localized on chromosome 2 in the vicinity of the NAT8 gene and may represent a pseudogene of NAT8. This gene contains two polymorphic nonsense mutations that disrupt the active site of the protein. The full-length product of this gene contains a complete acetyltransferase domain and is identical in length to NAT8.
Specifications
NAT8B | |
Polyclonal | |
Unconjugated | |
NAT8B | |
Acetyltransferase 1; ATase1; camello-like 1 like; camello-like protein 1; Camello-like protein 2; camello-like protein 6; Cml1; CML2; Cml6; Hcml2; N-acetyltransferase 8B; N-acetyltransferase 8B (GCN5-related, putative, gene/pseudogene); N-acetyltransferase 8B (putative, gene/pseudogene); N-acetyltransferase Camello 2; Nat8; Nat8b; NAT8BP; probable N-acetyltransferase 8B; probable N-acetyltransferase CML6; Putative N-acetyltransferase 8B; RGD621605 | |
Rabbit | |
Affinity chromatography | |
RUO | |
171084, 434057, 51471 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E0CYC6, Q9UHF3 | |
NAT8B, Nat8b-ps | |
Recombinant fusion protein containing a sequence corresponding to amino acids 75-165 of human NAT8B (NP_0574312). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.