Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT8L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537999
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537999 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537999 Supplier Invitrogen™ Supplier No. PA537999
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with human, rat, dog and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

This gene encodes a single-pass membrane protein, which contains a conserved sequence of the GCN5 or NAT superfamily of N-acetyltransferases and is a member of the N-acyltransferase (NAT) superfamily. This protein is a neuron-specific protein and is the N-acetylaspartate (NAA) biosynthetic enzyme, catalyzing the NAA synthesis from L-aspartate and acetyl-CoA. NAA is a major storage and transport form of acetyl coenzyme A specific to the nervous system. The gene mutation results in primary NAA deficiency (hypoacetylaspartia). [provided by RefSeq, Dec 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT8L
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NAT8L
Gene Accession No. Q3UGX3, Q8N9F0
Gene Alias 1110038O08Rik; 2.3.1.17; Camello-like protein 3; CML3; NAA synthetase; NACED; N-acetylaspartate synthetase; N-acetyltransferase 8 like; N-acetyltransferase 8-like; N-acetyltransferase 8-like (GCN5-related, putative); N-acetyltransferase 8-like protein; NAT8L; NAT8-LIKE; Protein Shati; RGD1305719; Shati
Gene Symbols NAT8L
Host Species Goat
Immunogen Peptide with sequence C-SVDSRFRGKGIAK, from the internal region of mouse Nat8l (aa211-223).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269642, 339983
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.