Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAT8L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568424
Description
Predicted to react with Mouse and Rat samples.
This gene encodes a single-pass membrane protein, which contains a conserved sequence of the GCN5 or NAT superfamily of N-acetyltransferases and is a member of the N-acyltransferase (NAT) superfamily. This protein is a neuron-specific protein and is the N-acetylaspartate (NAA) biosynthetic enzyme, catalyzing the NAA synthesis from L-aspartate and acetyl-CoA. NAA is a major storage and transport form of acetyl coenzyme A specific to the nervous system. The gene mutation results in primary NAA deficiency (hypoacetylaspartia). [provided by RefSeq, Dec 2010].
Specifications
NAT8L | |
Polyclonal | |
Unconjugated | |
NAT8L | |
1110038O08Rik; 2.3.1.17; Camello-like protein 3; CML3; NAA synthetase; NACED; N-acetylaspartate synthetase; N-acetyltransferase 8 like; N-acetyltransferase 8-like; N-acetyltransferase 8-like (GCN5-related, putative); N-acetyltransferase 8-like protein; NAT8L; NAT8-LIKE; Protein Shati; RGD1305719; Shati | |
Rabbit | |
affinity chromatography | |
RUO | |
269642, 289727, 339983 | |
-20°C | |
Liquid |
Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
D3ZVU9, Q3UGX3, Q8N9F0 | |
NAT8L | |
Synthetic peptide corresponding to residues near the C terminal of human NAT8L. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction