Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAT8L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583508
Description
Immunogen sequence: MADIEQYYMK PPGSCFWVAV LDGNVVGIVA ARAHEEDNTV ELLRMSVDSR FRGKGIAKAL GRKVLE.
This gene encodes a single-pass membrane protein, which contains a conserved sequence of the GCN5 or NAT superfamily of N-acetyltransferases and is a member of the N-acyltransferase (NAT) superfamily. This protein is a neuron-specific protein and is the N-acetylaspartate (NAA) biosynthetic enzyme, catalyzing the NAA synthesis from L-aspartate and acetyl-CoA. NAA is a major storage and transport form of acetyl coenzyme A specific to the nervous system. The gene mutation results in primary NAA deficiency (hypoacetylaspartia). [provided by RefSeq, Dec 2010].
Specifications
NAT8L | |
Polyclonal | |
Unconjugated | |
NAT8L | |
1110038O08Rik; 2.3.1.17; Camello-like protein 3; CML3; NAA synthetase; NACED; N-acetylaspartate synthetase; N-acetyltransferase 8 like; N-acetyltransferase 8-like; N-acetyltransferase 8-like (GCN5-related, putative); N-acetyltransferase 8-like protein; NAT8L; NAT8-LIKE; Protein Shati; RGD1305719; Shati | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339983 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N9F0 | |
NAT8L | |
Recombinant protein corresponding to Human NAT8L. Recombinant protein control fragment (Product #RP-96770). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction