Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAT9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554521 Supplier Invitrogen™ Supplier No. PA554521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRLNQNTLLL GKKVVLVPYT SEHVPRYHEW MKSEELQRLT ASEPLTLEQE YAMQCSWQED ADKCTFI Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.

NAT-9 (N-acetyltransferase 9), also known as EBSP (embryo brain-specific protein), is a 207 amino acid protein belonging to the acetyltransferase family and the GNAT subfamily. Containing a N-acetyltransferase domain, NAT-9 may be associated with psoriasis and psoriatic arthritis, a type of inflammatory/autoimmune disease that affects skin, tendons and/or joints of the hands and feet. Expressed as two isoforms produced by alternative splicing events, NAT-9 is encoded by a gene located on human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAT9
Gene Accession No. Q9BTE0
Gene Alias EBS; EBSP, hNATL; embryo brain specific protein; Embryo brain-specific protein; N-acetyltransferase 9; N-acetyltransferase 9 (GCN5-related, putative); N-acetyltransferase 9 (putative); NAT9
Gene Symbols NAT9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NAT9. Recombinant protein control fragment (Product #RP-93220).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.