Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Natamycin, 95%

Click to view available options
Quantity:
50mg
100mg
7681-93-8
C33H47NO13
665.733
MFCD00135085
NCXMLFZGDNKEPB-UWSQUGIJSA-N
129317863
CC1CC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(CC3C(O3)C=CC(=O)O1)O)O)O)C(=O)O)OC4C(C(C(C(O4)C)O)N)O
7681-93-8
C33H47NO13
665.733
MFCD00135085
NCXMLFZGDNKEPB-UWSQUGIJSA-N
129317863
CC1CC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(CC3C(O3)C=CC(=O)O1)O)O)O)C(=O)O)OC4C(C(C(C(O4)C)O)N)O
Applications
Pimaricin is an amphoteric antibiotic from Streptomyces chattanoogensis. It is used as an ergosterol and cholesterol binding agent to study lipid bilayer dynamics. It is used to study the pleitropic regulator AdpAch and the gamma-butyrolactone regulatory system of Streptomyces chattanoogensis.

Solubility
Soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Merck Index 14,6427
Solubility Information Soluble in DMSO
Formula Weight 665.72
Physical Form Powder
Percent Purity 95%
Boiling Point 952°C
Color White
Melting Point 280°C
Quantity 50mg
Sensitivity Light Sensitive
Chemical Name or Material Natamycin
Show More Show Less

RTECSNumber : TK3325000

storageNote1 : 2° to 8°C