Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Natamycin, 95%

Catalog No. p-7057503
Click to view available options
Quantity:
50 mg
100 mg

Natamycin (pimaricin, CAS # 7681-93-8) is a polyene macrolide antibiotic with antifungal properties. | CAS: 7681-93-8 | C33H47NO13 | 665.733 g/mol

Pimaricin is an amphoteric antibiotic from Streptomyces chattanoogensis. It is used as an ergosterol and cholesterol binding agent to study lipid bilayer dynamics. It is used to study the pleitropic regulator AdpAch and the gamma-butyrolactone regulatory system of Streptomyces chattanoogensis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Natamycin is a member of the macrolide antibiotic class, compounds originally derived from Saccharopolyspora erythraea, a type of soil bacteria

• Natamycin is an amphoteric compound with both acid and basic properties

• Natamycin exhibits antifungal activity • Natamycin exerts its effects by binding sterols within fungal cell walls and increasing membrane permeability, leading to a loss of homeostasis within the cell

Application

• Natamycin is suitable for use in the laboratory study of fungal keratitis and other infections of fungal origin

• Natamycin may also be used in laboratory research of cell physiology and mechanisms of antibiotic resistance

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-93-8
Molecular Formula C33H47NO13
Molecular Weight (g/mol) 665.733
MDL Number MFCD00135085
InChI Key NCXMLFZGDNKEPB-UWSQUGIJSA-N
PubChem CID 129317863
SMILES CC1CC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(CC3C(O3)C=CC(=O)O1)O)O)O)C(=O)O)OC4C(C(C(C(O4)C)O)N)O

Specifications

Type Natamycin
Physical Form Powder
Melting Point 280°C
Color White
Quantity 50 mg
Flash Point >110°C (230°F)
Sensitivity Light sensitive
Boiling Point 952°C
Merck Index 14,6427
Solubility Information Soluble in DMSO
Formula Weight 665.72
Percent Purity 95%
Chemical Name or Material Natamycin
Show More Show Less
EINECSNumber 231-683-5
RTECSNumber TK3325000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.