Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nateglinide
GREENER_CHOICE

Catalog No. 423110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-311-0 10 mg
42-315-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-311-0 Supplier Tocris Bioscience™ Supplier No. 4231/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Kir6 (KATP) blocker; displays high affinity for SUR1/Kir6.2 channels

Kir6 (KATP) blocker. Exhibits in vitro tissue selectivity for pancreatic β-cell-type Kir6 channels over cardiovascular Kir6 channels; displays high affinity for SUR1/Kir6.2 channels. Hypoglycemic agent; stimulates insulin secretion from pancreatic β-cells by increasing cytosolic Ca2+ concentration. Also agonist at the orphan receptor MRGPRX4.

Chemical Identifiers

CAS 105816-04-4
Molecular Formula C19H27NO3

Specifications

Quantity 10 mg
Formula Weight 317.42
Purity >98%
Molecular Weight (g/mol) 317.429
Chemical Name or Material Nateglinide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.