Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Assembled Target™ 10-425 Caps and Septa

Catalog No. p-4526253
Click to view available options
Color:
Natural
Septum:
Integral Molded Polypropylene

Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.

Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.

  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Storage cap has a solid top and pre-inserted PTFE disk liner
TRUSTED_SUSTAINABILITY

Specifications

Description Natural; Septa: Integral Molded Polypropylene
Color Natural
Material Polypropylene
Septum Integral Molded Polypropylene
Product Line National Assembled Target
Type Screw Cap
Closure Size 10 mm
For Use With (Equipment) 10-425 Screw Thread Vials

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.