Learn More
Thermo Scientific™ National Economy Vials
Description
Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.
Specifications
Specifications
| Description | 60180-702 |
| Color | Amber |
| Volume (Metric) | 2 mL |
| Volume (Metric) Working | 2 mL |
| For Use With (Equipment) | Thermo Scientific, Agilent, PE, Shimadzu, Varian |
| Product Type | Crimp Top Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.